Learn More
AirClean™ Systems Bonded Carbon Filters
AirClean™ Systems Bonded Carbon Filters used in evidence drying cabinets to absorb the fumes and odors.
Supplier: AirClean™ Systems ACF20918
Description
- Used in evidence drying cabinets to absorb fumes and odors associated with drying and decay of bloody articles
Specifications
ACF209-18 | |
15 lb. | |
30.4 x 45.7 x 7.62 cm (12 x 18 x 3 in.) | |
For Inorganics |
AC648 Units with Negative Pressure | |
6.8 Kg | |
Filter |