Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ahlstrom™ Filter Papers – Grade 238 PROMO
Specifications
For Use With (Application) | Chromatography applications. Use as wicks in protein or nucleic acid blotting. Suited for electrophoresis |
---|---|
Grade | 238 |
Thickness (Metric) | 0.35 mm |
Catalog Number | Mfr. No. | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
22279539PM
|
Ahlstrom
23884657PM |
46 cm | 57 cm | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
0979053BPM
|
Ahlstrom
23880710PM |
7 cm | 10 cm | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
0979053GPM
|
Ahlstrom
23882020PM |
20 cm | 20 cm | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Specifications
Chromatography applications. Use as wicks in protein or nucleic acid blotting. Suited for electrophoresis | |
238 | |
0.35 mm |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title