Learn More
ASSOCIATED METALS AGFA FULL-SPD BLU 8X10 FLM 100
Supplier: ASSOCIATED METALS UPB0810
Dimensions : 20.3 X 25.4 Cm (8 X 10") Sensitivity : Blue Speed : 800 Supplier No. : Upb0810 Unit : Case Of 5 High Quality General Radiographic Film May Be Used With Any Medical X-Ray Machine For Routine X-Rays In Primary Medical Procedures. Definition Is Medium Contrast. Blue Sensitive Film Is Available In High Speed 800 Or Half Speed 400. Speeds Are Dependent Upon Screen Being Used. Store In Cool, Dry Place.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.