Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Agarose NA Media
Highly purified agarose for separation of nucleic acids to be used as substrates for enzymes. Cytiva™ Agarose NA Media with low electroendosmosis (EEO) is recommended for most DNA/RNA applications.
$405.00
Specifications
Color | White |
---|---|
Product Type | Agarose |
Form | Solid |
Material | Agarose |
Melting Temperature | 60° to 90°C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
45-000-100
|
Cytiva
17055402 |
100 g |
Each of 1 for $405.00
|
|
|||||
Description
- Gelling temp 34 to 37°C
Specifications
White | |
Solid | |
60° to 90°C |
Agarose | |
Agarose |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title