Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Agarose 2%
SCI_ED

Catalog Number S25130
Click to view available options
Quantity:
200 mL
Packaging:
2%
9012-36-6
Powder
9012-36-6
Powder

This product is an Educational Grade Chemical — no C of A is available.

Quantity 200 mL
Packaging 2%
Chemical Name or Material Agarose
ShelfLife 2 years
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit