Learn More
Thermo Scientific AG11 ANION GUARD COLUMNS
Supplier: Thermo Scientific 044078
IonPac AG11 Guard Column, Size:4 mmx 50 mm, Provides fast analysis of inorganic anions and organic acid anions, including polyphosphates, using hydroxide gradient elution