Learn More
Ted Pella Inc Highest Grade V1 AFM Mica Discs, 10mm, 10/pk
Supplier: Ted Pella Inc 50
Highest quality grade V1 mica, 0.21mm (0.0085”) thick. Interleaved, in packages of 10. Can be mounted on AFM/STM discs.