Learn More
Thermo Scientific AEROSALT NEBULISER
Supplier: Thermo Scientific 842312051331
AeroSalt Nebuliser; Nebuliser intended for the analysis of samples containing relatively; high solids (up to 15%).; Compatible with all iCAP 6000 Series Spectrometers