Learn More
Advantus OpticPad™ Optical Surface Cleaning Pad
Click to view available options
Box of 100
Description
Ideal for cleaning and polishing mirrors, microscope lenses, machine optics, magnifying glasses, optical sensors, safety goggles and fiber optic splice connections
- Nonabrasive
- Individually wrapped
- Disposable, lint-free pad pre-saturated with solvent for cleaning and polishing optical-grade surfaces
Specifications
Product Type | Optical Surface Cleaning Pad |
For Use With (Application) | Cleaning and polishing front and back surface mirrors, microscope lenses, machine optics, magnifying Glasses, optical sensors and safety goggles, cleaning Fiber optic splice connections |