Learn More
Advantech Fines Collector for Model L3P Sonic Sifter Separators
For use with L3P sonic sifter separators
Supplier: Advantech L3N5
Description
For use with L3P sonic sifter separators
- Fines Collector stops dust at bottom of air column in sifter and transmits oscillary motion to air column
Specifications
Fines Collector | |
Stops dust at bottom of sifter air column, transmits oscillary motion to air column | |
L3P sonic sifter separators |