Learn More
MilliporeSigma™ Supelco™ Adsorbent Scrapers
For Rapid Removal of Adsorbent from TLC Plates
Supplier: MilliporeSigma™ Supelco™ Z2652761PAK
Description
- Flat 13 mm steel blade tapered to a sharp edge
- Ideal for rapid removal of adsorbent from TLC plates
- Aluminum handle
- Replaceable blade
Specifications
5/Pk. | |
Replacement Blades | |
Rapid Removal of Adsorbent From TLC Plates | |
Adsorbent Scrapers |