Learn More
Clontech Labs 3P Adeno-X™ qPCR Titration Kit
Supplier: Clontech Labs 3P 632252
The Adeno-X qPCR Titration Kit is designed to be used with real-time PCR instruments to rapidly titrate adenoviral stocks from all Ad5-based adenoviral vectors. The protocol combines qPCR with green intercalating dye chemistry, allowing you to determine the viral genome copy number in adenoviral preparations (i.e., crude lysates or purified stocks) from a calibrated DNA standard curve. The kit includes a viral nucleic acid purification kit, a calibrated Adeno-X DNA Control Template, primers, and all the reagents required to conduct 200 TB Green-based qPCR reactions with either the DNA Control Template or purified viral DNA.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.