Learn More
Clontech Labs 3P ADENO-X GOSTIX
Supplier: Clontech Labs 3P 632270
Adenovirus titration kits accurately measure functional titer of your adenovirus preparation using a hexon antibody, or by SYBR Green-based, real-time qPCR in which the virus sampleAEs Ct is compared to a standard curve.