Learn More
Chemglass Life Sciences 50mL Addition Funnel, 14/20 Joint Size, 2mm PTFE Stopcock, 220mm OAH
Supplier: Chemglass Life Sciences CG170214
50mL Addition Funnel, 14/20 Joint Size, 2mm PTFE Stopcock, 220mm OAH. Cylindrical style addition funnel with a standard taper top outer joint and lower inner joint, both being the same size. With pressure equalizing arm and PTFE stopcock. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.