Learn More
Chemglass Life Sciences 250mL Funnel, Addition, 24/40 Joint Size, Valve On Side Arm, Airfree, Schlenk
Supplier: Chemglass Life Sciences AF054821
250mL Funnel, Addition, 24/40 Joint Size, Valve On Side Arm, Airfree, Schlenk. Lower outlet is a standard taper inner joint and has a CG-961-01 0-4mm Chem-Vac™ Chem-Cap valve. Top outer joint is the same size as the lower inner joint. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retaining device or a 0-4mm Chem-Cap Airfree inlet valve. This item comes with the inlet valve.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.