Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Adapters for Double Blood Bag Buckets for Thermo Scientific™ HAEMAFlex™ 12/16 Rotors
Select adapters for double quint blood bags or hook adapters for cord blood separations in double blood bag buckets for Thermo Scientific™ HAEMAFlex™ 12/16 rotors.
$265.50 - $354.00
Catalog Number | Mfr. No. | Description | Capacity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Capacity | Price | Quantity & Availability | ||||
75-003-851
|
Thermo Scientific™
75003851 |
Double Quint Blood Bag Adapter for XXL size (110 x 88mm) (set of 2) | 12/16 x 500 mL |
Pack of 2 for $265.50
|
|
||||
75-003-852
|
Thermo Scientific™
75003852 |
Double Quint Blood Bag Adapter for XL size (110 x 76mm) (set of 2) | 12/16 x 500 mL |
Pack of 2 for $265.50
|
|
||||
75-003-853
|
Thermo Scientific™
75003853 |
Double Quint Blood Bag Adapter for M size (110 x 57mm) (set of 2) | 12/16 x 450 mL |
Pack of 2 for $265.50
|
|
||||
75-003-855
|
Thermo Scientific™
75003855 |
Hook Adapter for Cord Blood Separations, for use with M size Adapters (set of 2) | 12/16 x 300 mL |
Each for $336.50
|
|
||||
75-003-868
|
Thermo Scientific™
75003868 |
Hook Adapter for Cord Blood Separations, for use with XXL size Adapters (set of 2) | 12/16 x 300 mL |
Each for $354.00
|
|
||||
Description
Thermo Scientific blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.
For high-throughput blood processing needs, the HAEMAFlex 12 swinging bucket rotor provides 12 x 500mL capacity or upgrade your centrifuge capacity to 16 x 500mL blood bags with the HAEMAFlex 16 rotor and the same bucket selections.Select adapters for double quint blood bags (XXL, XL or M size) or hook adapters for cord blood separations (XXL or M size).
Specifications
Double Blood Bag Bucket |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title