Learn More
Eppendorf™ Adapter for S-4xUniversal Rotor
The Adapter for S-4xUniversal Rotor facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for the S-4xUniversal Rotor.
$424.50 - $675.50
Specifications
For Use With (Equipment) S-4xUniversal Rotor
No. per Pack 2
View More Specs
Products 12
Catalog Number Mfr. No. Capacity Tube Type No. of Places Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Tube Type No. of Places Price Quantity & Availability  
05-414-351
View Documents
Eppendorf™
5910764006
7 x 50 mL Skirted Tubes, 7 x 30 mL Oak Ridge Tubes Skirted Bottom, Oak Ridge 7/7
Pack of 2 for $424.50
Add to Cart
13-864-449
View Documents
Eppendorf™
5910769008
9 x 50 mL Conical Tubes, 1 Microplate Conical 9/1
Each for $523.00
Add to Cart
05-414-353
View Documents
Eppendorf™
5920700006
1 x 1 L Nalgene Bottle Flat Bottom 1
Pack of 2 for $427.50
Add to Cart
13-864-451
View Documents
Eppendorf™
5910760000
1 x 500 mL Conical Bottle Conical 1
Each for $480.50
Add to Cart
13-864-452
View Documents
Eppendorf™
5910757000
1 x 750 mL Flat-bottom Bottle Flat Bottom 1
Each for $481.50
Add to Cart
13-864-453
View Documents
Eppendorf™
5910753004
5 x MTP/DWP/PCR Plates PCR 5
Each for $480.50
Add to Cart
13-864-450
View Documents
Eppendorf™
5910751001
7 x 50 mL Conical Tubes, 1 x 96/384-well Microplate, 1 x 250 mL Flat-bottom Bottle, 1 x 175 to 225 mL Conical Bottle Flat/Conical Bottom 7/1/1/1
Each for $675.50
Add to Cart
05-414-350
View Documents
Eppendorf™
5910758006
46 tubes of 12 x 75mm or 5ml, Round Bottom Tubes (FACS) Round Bottom (FACS) 46
Pack of 2 for $437.00
Add to Cart
13-864-448
View Documents
Eppendorf™
5910752008
16 x 5 mL Microtubes, 16 x 15 mL Conical Tubes, 1 MTP/DWP Microtube, Conical 16/16/1
Each for $480.50
Add to Cart
13-864-447
View Documents
Eppendorf™
5910762003
21 x 9 mL Round Bottom Tubes Round Bottom 21
Each for $424.50
Add to Cart
13-864-446
View Documents
Eppendorf™
5910754000
26 x 7.5 to 12 mL Round Bottom Tubes Round Bottom 26
Each for $481.50
Add to Cart
13-864-445
View Documents
Eppendorf™
5910755007
30 x 4 to 8 mL Round Bottom Tubes Round Bottom 30
Each for $480.50
Add to Cart
Description
Description
  • Suitable for S-4xUniversal rotor buckets
  • Accommodates selected array of tubes, bottles, or plates
Specifications
Specifications
S-4xUniversal Rotor
2