Learn More
Ace Glass, Inc. Adapter, filter, PTFE, #80 Ace-Thred to 3/8in female NPT, 100micron polyethylene filter, 350 micron polypropylene filter, glass filter support, retaining ring, CAPFE O-ring
Supplier: Ace Glass, Inc. 586107
Adapter, filter, PTFE, #80 Ace-Thred to 3/8in female NPT, 100 micron polyethylene filter, 350 micron polypropylene filter, glass filter support, retaining ring, CAPFE O-ring.