Learn More
Chemglass Life Sciences Adapter, Distillation, 0-14mm Valve with 45/50 Lower Inner Jt
Supplier: Chemglass Life Sciences CG1965D210
Adapter, Distillation, 0-14mm Valve with 45/50 Lower Inner Jt. Part of Chemglass distillation kit for reactor systems. See CG-1965-D-110 and CG-1965-D-120 for full distillation kits.