Learn More
Health Care Logistics Inc ADAPTER CAPS, 20MM/B 20/PK
SDP
Supplier:  Health Care Logistics Inc 801210
Encompass_Preferred
Easy to Use - Converts a standard medication bottle into a filling device for plastic oral dispensers.Let Us Help - To match the right adapter cap to a bottle, request sizing template from Customer Service or print the template by clicking on the Adapta-Cap Icon below (print at 100% for the most accurate sizing).Material - Low-density polyethyleneSize - 20mm/BQuantity - 20 per package
Catalog No. NC1862557