Learn More
Acuderm Inc ACU-PUNCH 7MM
Supplier: Acuderm Inc P750
Acu-Punch; 7mm; Acuderm;; Sterile; Disposable Skin Biopsy Punch; Only punch available in a complete range of 14 sizes from 1 to 12mm; Razor sharp and ready to use; Handle is ribbed for maximum control even when wearing gloves; 50/BX