Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile (Certified), Fisher Chemical

Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
75-05-8
C2H3N
41.053
MFCD00001878
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
6342
CHEBI:38472
acetonitrile
CC#N
75-05-8
C2H3N
41.053
MFCD00001878
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
6342
CHEBI:38472
acetonitrile
CC#N

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Melting Point -45°C
Assay Percent Range ≥98.5 %
Linear Formula CH3CN
UN Number 1648; Colorless
Formula Weight 41.05
Percent Purity ≥99.9%
Grade Certified
Density 0.78g/mL
Boiling Point 81.6°C
Quantity 500 mL
Flash Point 1°C (33°F)
Identification Pass Test
Packaging Glass Bottle
Water 0.03% max.
Chemical Name or Material Acetonitrile
Show More Show Less

DOTInformation : Class 3: Flammable Liquid

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit