Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetonitrile Anhydrous (Septum-Sealed), Fisher BioReagents
Click to view available options
Quantity:
50 mL
Packaging:
Glass Serum
Description
Septum-sealed acetonitrile is used in critical applications that require the complete absence of water.
Specifications
Melting Point | -46°C |
Absorbance | 0.01 AU max. at 254nm, 0.02 AU max. at 220nm, 0.04 AU max. at 210nm, 0.05 AU max. at 205nm, 0.07 AU max. at 200nm, 0.15 AU max. at 195nm, 100 AU max. at 190nm |
Color | Colorless |
Decomposition Information | >500°C |
Density | 0.781g/cm³ |
Boiling Point | 81°C |
Quantity | 50 mL |
Packaging | Glass Serum |
Physical Form | Liquid |
Refractive Index | 1.3405 to 1.3425 (at 25°C) |
Show More |
Safety and Handling
Recommended Storage | RT |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction