Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetaminophen, Granular, USP, 98-102%, Spectrum™ Chemical

Click to view available options
Quantity:
12 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
Description
Spectrum™ Chemical Acetaminophen, Granular, USP , also known as paracetamol, is a widely used antipyretic and mild analgesic that is available over-the-counter. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 100% |
Ignition Residue | 0.1% |
Loss on Drying | 0.5% |
Packaging | Fiber Drum |
Quantity | 50 kg |
Percent Purity | 98 to 102% |
Grade | USP |