Learn More
JANT PHARMACAL CORPORATION Accutest URS-10 Urine Reagent Strips
SDP
Supplier:  JANT PHARMACAL CORPORATION UA710A
Encompass_Preferred
Accutest URS-10 Urine Reagent Strips for Urinalysis are in vitro diagnostic test devices that use reagents for qualitative & semi-quantitative urinalysis. Accutest URS-10 Urine Reagent Strips are for single use in professional near patient (point-of-care) facilities & centralized laboratory locations by medical tech. Benefits & Features: Accutest URS-10 Urine Reagent Strips for Urinalysis are intended for use to detect conditions indicating possible diabetes, metabolic abnormalities, liver diseases, kidney function, & urinary tract infections. Test results can be used along with other diagnostic information to rule out certain disease states & to determine if microscopic analysis is needed. High Quality Test Strips; Fast Qualitative Results; Reliable Sensitivity; Easy to Read; CLIA WAIVED when read visually; May be used on Siemens CLINITEK Analyzers; 10 Parameter: Leukocytes, Nitrite, Urobilinogen, Protein, pH, Blood, Specific Gravity, Ketone, Bilirubin, Glucose.
Catalog No. NC0985300
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.