Learn More
Quip Laboratories Inc AccuPoint Advanced HC System
SDP
Supplier:  Quip Laboratories Inc QLI9903HCQ
Encompass_Preferred
The AccuPointAdvanced HC System is an ATP cleaning verification and tracking system that measures cleaning effectiveness. The system features a hand-held instrument and sampling device that measure ATP levels on a variety of surfaces, including liquid containers, dispensers, receptacles and automated systems.
Catalog No. NC1828109