Learn More
Fisherbrand™ accumet™ Basic AB315 Benchtop Laboratory pH/mV Meter PROMO
Ideal for daily pH testing of your routine laboratory samples, the accumet™ Basic AB315 Benchtop Meter provides essential functions needed for common pH measurements without complicated functions, all at a budget-friendly price.
$606.00 - $689.00
Specifications
Accuracy (mV) | ±0.2 or ±0.05% |
---|---|
Accuracy (pH) | ±0.01 |
Accuracy (Temperature) | ±0.3°C (±0.5°F) |
Data Management Details | Printed or CSV Format |
Outputs | Port for Computer or Printer |
Catalog Number | Includes | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Includes | Price | Quantity & Availability | ||||||
36-AB3-15AS23
|
pH/ATC Epoxy-body Double Junction Gel-filled Electrode, Electrode Stand, Power Adapter |
Each for $606.00
|
|
||||||
36-AB3-15BS23
|
Glass-body Double Junction Refillable pH Electrode, ATC Probe, Electrode Stand, Power Adapter |
Each for $689.00
|
|
||||||
Description
- Large, backlit display simultaneous shows pH, mV and temperature readings
- One to five point pH calibration with automatic buffer recognition of four pH buffer sets
- One point relative mV calibration (RmV) for ORP measurements
- One point ATC temperature probe calibration
- Calibration due alarm, selectable 1 to 60 hr., or off
- Automatic or manual temperature compensation
- Ready icon indicates measurement stability
- Electrode condition icon indicates electrode status 500 data set storage with time/date stamp
- Non-volatile meter memory preserves settings and data in event of power loss
- Meter-attached stand facilitates easy electrode movement into and out of samples
Specifications
±0.2 or ±0.05% | |
±0.3°C (±0.5°F) | |
Port for Computer or Printer | |
pH, Relative mV, Temperature | |
0.01, 0.1 | |
±2000 mV | |
1 to 5 (pH), 1 (Relative mV), 1 (Temperature) | |
23°F to 221°F | |
5 in. LCD | |
Non-volatile | |
Automatic, Manual | |
CE, TUV 3-1, FCC A, Without NIST-traceable pH Certificate | |
Universal Power Adapter | |
AB315 | |
Indicator Icons | |
1 Channel | |
50/60 Hz |
±0.01 | |
Printed or CSV Format | |
pH with mV and Temperature, Relative mV with mV and Temperature | |
-2 to 18 | |
USA: 2, 4.01, 7, 10.01, 12; NIST: 1.68, 4.01, 6.87, 9.18, 12.46; DIN: 1.09, 3.06, 4.65, 6.79, 9.23, 12.75; FSCI: 1, 3, 6, 8, 10, 13 | |
1 mV, 0.1 RmV | |
-5°C to +105°C | |
0.1°C (0.1°F) | |
Yes | |
500 Points | |
BNC, ATC | |
IP54 | |
3 Years | |
±2000 mV | |
Automatic, Manual | |
100/240 V |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.