Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform Signs Marking Paints
Mark sites and locations as well as stripe underground lines, aisles, pathways, parking lines and traffic lines
Specifications
Quantity | 1 Ea. |
---|---|
Capacity (English) | 17 oz. |
Product Type | Paint |
For Use With (Application) | Solvent based Utility, Mark Sites and Stripe Underground Lines, Aisles, Pathways, Parking, Traffic Lines |
Certifications/Compliance | VOC |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
19-804-454
|
Accuform Signs
FMP229 |
Yellow | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
19-821-767
|
Accuform Signs
FMP210 |
Fluorescent Orange | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Solvent based
- Can be applied to virtually any indoor and outdoor surface (concrete, asphalt, gravel, soil and grass)
- 17 oz. cans are designed for upside-down use with non-clogging spray tips
- Contains fast-drying, long-lasting, high-solids formula paint that penetrates and adheres quickly to most surfaces
- Lead-free
- Available in a variety of colors
Not available for sale in Canada
Order Info
Due to DOT regulations this item must ship via ground service only. It is not eligible to ship overnight, 2-day or 3-day.
Compliance
VOC compliant
Specifications
1 Ea. | |
Paint | |
VOC |
17 oz. | |
Solvent based Utility, Mark Sites and Stripe Underground Lines, Aisles, Pathways, Parking, Traffic Lines |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title