Learn More
Roche Diagnostics ACCU-CHEK Safe-T-Pro Uno Lancets
Designed to reduce the spread of infection and meet the latest safety requirements while minimizing pain for patients. Roche Diagnostics Accu-Chek Safe-T-Pro Uno Lancets are suitable for all blood sample needs.
Supplier: Roche Diagnostics 05888662160
Description
Each Accu-Chek lancet has a retractable needle designed to reduce the spread of infection and meet the latest safety requirements.- 1.5 mm depth setting
- 28 g retractable needle
- Approved for use with neonates
- Release button
- Sterility cap
Specifications
Safe-T-Pro Uno | |
Blood sampling | |
White and Yellow | |
1.5mm |
28g | |
Retractable needle | |
Yes |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.