Learn More
Fisherbrand™ Absorbent Underpads PROMO
These Absorbent Underpads quickly absorb liquids and keep spills from spreading.
Catalog No. 14-206-62AC22
Add to cart
This item is not returnable. View return policy
Description
Description
  • Cushioned surface minimizes breakage of delicate labware
  • Made with high-absorbency fibers
  • Polyethylene backing is chemical resistant, and the spunbond top layer provides durability even when wet
  • Lie flat, hugging the benchtop to prevent sliding
  • Absorb up to 750 mL/m2
Specifications
Specifications
Absorbent Underpad
750 mL/m2
610 mm
510 mm
50 Pk.
Polyethylene
24 in.
20 in.
Quickly absorb liquids and keep spills from spreading
Mat