Learn More
Vector Laboratories VECTASTAIN Elite ABC Universal PLUS Kit, Peroxidase (Horse Anti-Mouse/Rabbit IgG)
Supplier: Vector Laboratories PK8200
The VECTASTAIN Elite ABC system is the most sensitive avidin/biotin-based peroxidase system and is approximately 5 times more sensitive than the original VECTASTAIN ABC Kit with the same low background. Our Universal Biotinylated Anti-Mouse/Rabbit IgG secondary antibody included in this kit is a cocktail of biotinylated anti-mouse IgG and anti-rabbit IgG, designed for use with both rabbit and mouse primary antibodies. This antibody should not be used to stain tissues from rodents or rabbits due to its reactivity with IgG present in the tissues of these species.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.