Learn More
Vigene Biosciences Inc AAV8 STNRD(FULL CAPSIDS)100UL
Manufacturer:  Vigene Biosciences Inc RSAAV8FL
Encompass
AAV8 reference standards (full capsids), 7.97x10^11 GC/mL, 100 uL
Catalog No. NC1875233