Learn More
Vigene Biosciences Inc AAV6 REFERENCE STANDARDS,100UL
Manufacturer:  Vigene Biosciences Inc RSAAV6ET
Encompass
AAV6 reference standards (empty capsids), 1.79x10^12 VP/mL, 100 ul
Catalog No. NC1875606