Learn More
Vigene Biosciences Inc AAV5 STNRD(FULL CAPSIDS)100UL
Manufacturer:  Vigene Biosciences Inc RSAAV5FL
Encompass
AAV5 reference standards (full capsids), 2.60x10^11 GC/mL, 100 uL
Catalog No. NC1875231