Learn More
Vigene Biosciences Inc AAV5 REFERENCE STANDARDS,100UL
Manufacturer:  Vigene Biosciences Inc RSAAV5ET
Encompass
AAV5 reference standards (empty capsids), 1.46x10^12 VP/mL, 100 uL
Catalog No. NC1875604