Learn More
Vigene Biosciences Inc AAV2 REFERENCE STANDARDS,100UL
Manufacturer:  Vigene Biosciences Inc RSAAV2ET
Encompass
AAV2 reference standards (empty capsids), 1.27x10^12 VP/mL, 100 uL
Catalog No. NC1875602