Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kimberly-Clark Professional™ KleenGuard™ A30 Breathable Splash and Particle Protection Coveralls
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
8X-Large
Description
- Allows for maximum productivity and safety for you and your employees
- The material is strong and abrasion resistant, and filters out many water-based liquids and dry particles
- Includes seamless front (for primary exposure area), a 1 inch zipper flap, hood, elastic wrists and new skid-resistant boots that provide up to 68% improvement in traction on wet and dry surfaces when compared to our former boot material
- Protect against hazardous dry particles, aerosols and non-hazardous light liquid splashes, and they're perfect for environments such as pharmaceutical research, aerospace, maintenance and general manufacturing
Specifications
Size | Small |
Gender | Unisex |
Color | White |
Cuff Style | Elastic |
Closure Type | Zipper |
Coating | Anti-static |
Back Style | Elastic |
Certifications/Compliance | NFPA 99 Compliant |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction