Learn More
HARRIS PRODUCTS GROUP 9296NC-15-320 C02 GAS REGULATO
Supplier: HARRIS PRODUCTS GROUP 3302669
The Model 9296 Multi-StageTM regulator is designed for applications where a constant delivery pressure is required over a wide range of inlet pressures. A standard for general purpose laboratory, industrial and welding applications where neoprene diaphrag