Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

91-SOW Surr Spike, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Manufacturer:  SPEX CertiPrep CLPPS91H

Catalog No. CLPPS91H


Add to cart

Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Decachlorobiphenyl2051-24-32000μg/mL
2,4,5,6-Tetrachloro-m-xylene877-09-81000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

91-SOW Surr Spike
67-64-1, 2051-24-3, 877-09-8
Decachlorobiphenyl (2000μg/mL),2,4
1mL
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
49.85, 0.2, 0.1
2mL amber prescored glass ampule
Hexane:Acetone
GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Safety and Handling

Safety and Handling

Highly flammable liquid and vapor. Toxic if inhaled. Causes damage to organs.

DOTInformation : Nonhazardous

Recommended Storage : Freezer

SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Product intended for laboratory use only