Learn More
Tritech Research Inc 90% Platinum, 10% Iridium, 3 ft
Supplier: Tritech Research Inc PT9010
This 90% Platinum, 10% Iridium Wire is the most commonly preferred material for making C. elegans "worm picks".