Learn More
Weber Scientific Weber DB Sterilized Pre-filled Dilution Bottles, Blue, 90 mL, Phosphate Buffer, 72/cs
SDP
Supplier:  Weber Scientific 312759
Encompass
Phosphate Buffer for water and wastewater (APHA or EPA methods). Contains magnesium chloride, monobasic potassium phosphate and sodium hydroxide for pH adjustment.
Catalog No. NC1508154