Learn More
DANCO PACKAGING PRODUCTS LLC 8IN IMPULSE HEAT SEALER
Supplier:  DANCO PACKAGING PRODUCTS LLC 15MP8
Encompass_Preferred
Impulse Heat Sealer, 8 in. tabletop impulse heat sealer will seal polyethylene or polypropylene materials up to 5m in thickness, Creates a 1/16 in. seal.
Catalog No. NC9287857