Learn More
Metrohm USA 801 MAGNETIC STIRRER W/ STAND
Supplier: Metrohm USA 28010040
Magnetic stirrer including base plate, support rod and electrode holder for supplementing the Titrino plus, Dosimat plus, Titrandos, Sample Processors, 805 Dosimat and 780/781 pH meters as well as the 856 and 867 measuring modules. With permanently attached cable for MSB (Metrohm Serial Bus).
This item is not returnable.
View return policy