Learn More
BioSpec Products Vial Rack, Polycarbonate, for 7 ml screw-cap tubes
SDP
Supplier:  BioSpec Products 504VH12
Encompass_Preferred
Polycarbonate vial rack with a microplate footprint. Holds four to twelve 7 ml screw-cap vials (BioSpec's Cat No. 3205). Used for extraction and cell disruption of sample too large to be accommodated inside standard 2 ml microvials. Compatible with our MiniBeadbeater-96.
Catalog No. NC0997296
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.