Learn More
Cole-Parmer 50 Pound Solid Nylon Cable/Zip Ties, 7.5" L, White; 1000/Pk
Supplier: Cole-Parmer 0683056
These ties are injection-molded of 6/6 nylon, so they wont corrode or stretch. They have a durable locking mechanism that holds securely, even when used to bundle vibrating or flexing materials. These ties continue to hold tension even in oily spray or wet environments. Ties easily can be applied by hand or using a tensioning tool. Tools can be purchased separately.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.