Learn More
Malvern Panalytical 60Nm Polystyrene Latex
Supplier:  Malvern Panalytical LTX3060A
Encompass
Nanosphere Size Standards are highly uniform polystyrene spheres calibrated by NIST traceable standards and packaged as aqueous suspensions in dropper-tipped bottles. 60nm /15ml. “Nanosphere Size Standards are highly uniform polystyrene spheres calibrated by NIST traceable standards and packaged as aqueous suspensions in dropper-tipped bottles. 1% solids, optimized for easy dispersal and colloidal stability. Includes Certificate of Calibration and Traceability to NIST.”   
Catalog No. NC1152633