Learn More
Chemglass Life Sciences 60mL Funnel, Addition, Graduated, 14/20 Joint Size, Airfree, Schlenk
Supplier: Chemglass Life Sciences AF0547G01
60mL Funnel, Addition, Graduated, 14/20 Joint Size, Airfree, Schlenk. Pressure equalizing addition funnel, with Graduations, and a glass T-Bore stopcock in the equalizing arm permitting the purging and isolation of the funnel contents for storage. Both top outer and lower inner joints are the same size. Lower stopcock is a glass straight bore stopcock. Both stopcocks are 2mm high vacuum glass stopcocks having a Stainless Steel constant pressure retention device.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.