Learn More
Cole-Parmer Fischer Technology FMP 40 Dualscope Coating Thickness Gauge
Supplier:  Cole-Parmer 5410012
Encompass_Preferred
These easy-to-operate, rugged instruments are suitable for virtually any coating thickness measurement requirement. With a wide assortment of plug-in type probes, these meters are designed for nondestructive coating thickness measurements according to the magnetic induction method. In addition to adding up to 100 user-created calibration applications, the spectrum of requirements that this series is capable of spans from extensive painted specimens, to other very specific and complex measuring challenges. Select appropriate probes based on your application requirements. Probe not included and must be purchased seperately.
Catalog No. NC0495214
This item has been discontinued and is no longer available. Please reach out to your sales representative.