Learn More
Wilmad Labglass 600ML LYOPHILIZATION FLASK
Supplier: Wilmad Labglass LG13101108
Freeze drying flasks for VirTis� lyophilizing equipment. Openings are extra-wide for efficient shell freezing and suited for bulky samples. Wall thickness of flask is designed for vacuum applications. Complete assembly consists of the flask, the silicone cover, filter and o-ring. 600ML