Learn More
US Plastic Corporation 6 Gallon Polyethylene Tank with Spigot
SDP
Supplier:  US Plastic Corporation 14875
Encompass
12" L x 12" W x 12" H. Available in a wide range of sizes, Tamco tanks can be used for many different processing applications. Smaller sizes are excellent for table top operations, such as photographic processing, etching printed circuits and treating small parts, while larger sizes are great for many chemical plating, processing and storage applications.
Catalog No. NC1752373